4-Phenyl-3-butyn-2-one 98%

Supplier: Thermo Scientific Chemicals
Warning

17133-0010 17133-0050
TS17133-0010EA 55 USD
TS17133-0010 TS17133-0050
4-Phenyl-3-butyn-2-one 98%
4-Phenyl-3-butyn-2-one
Formula: C₁₀H₈O
MW: 144.17 g/mol
Boiling Pt: 75…76 °C (1.1hPa)
Density: 0.99 g/cm³
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00008776
CAS Number: 1817-57-8

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR