4-Methoxysalicylaldehyde 99%

Supplier: Thermo Scientific Chemicals
Warning

17108-0050 17108-0250
TS17108-0050EA 76 USD
TS17108-0050 TS17108-0250
4-Methoxysalicylaldehyde 99%
4-Methoxysalicylaldehyde
Formula: C₈H₈O₃
Boiling Pt: 124 °C (12 mmHg)
Melting Pt: 40…43 °C
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00003327
CAS Number: 673-22-3
EINECS: 211-604-0

Order Now

Specification Test Results

Appearance White to beige to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 40 to 43 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR