4,4'-Dimethoxybenzil 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: Bis(4-methoxyphenyl)ethanedione, Anisil, p-Anisil, 1,2-Bis(4-methoxyphenyl)ethane-1,2-dione

17045-0050 17045-0250
TS17045-0050EA 53 USD
TS17045-0050 TS17045-0250
4,4'-Dimethoxybenzil 99+%
4,4'-Dimethoxybenzil
Formula: C₁₆H₁₄O₄
MW: 270.28 g/mol
Boiling Pt: 215…219 °C (3 mmHg)
Melting Pt: 131…134 °C
MDL Number: MFCD00008405
CAS Number: 1226-42-2
EINECS: 214-960-5

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 131 to 134 °C
GC ≥99.0 %
Residue after ignition ≤0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR