ω-Bromo-4-methylacetophenone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: α-Bromo-4-methylacetophenone, Bromomethyl p-methylphenyl ketone, 2-Bromo-4'-methylacetophenone, 2-Bromo-1-(p-tolyl)ethanone, 4-Methylphenacyl bromide

17039-0050 17039-0250 17039-1000
TS17039-0050EA 54 USD
TS17039-0050 TS17039-0250 TS17039-1000
ω-Bromo-4-methylacetophenone 97%
ω-Bromo-4-methylacetophenone
Formula: C₉H₉BrO
MW: 213.07 g/mol
Melting Pt: 52…53 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000203
CAS Number: 619-41-0
EINECS: 210-595-0

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or needles
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR