3',4'-Dimethoxyacetophenone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(3,4-Dimethoxyphenyl)ethan-1-one, 3,4-Dimethoxyacetophenone, 1-(3,4-Dimethoxyphenyl)ethanone

16885-0250 16885-1000 16885-5000
TS16885-0250EA 35 USD
TS16885-0250 TS16885-1000 TS16885-5000
3',4'-Dimethoxyacetophenone 98%
3',4'-Dimethoxyacetophenone
Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Boiling Pt: 286…288 °C (760 mmHg)
Melting Pt: 48…52 °C
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00008737
CAS Number: 1131-62-0
EINECS: 214-468-0

Order Now

Specification Test Results

Appearance Yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 48 to 52 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR