4-Fluoro-3-nitroaniline 99%

Supplier: Thermo Scientific Chemicals
Warning

16840-0050 16840-0250 16840-1000
TS16840-0050EA 18 USD
TS16840-0050 TS16840-0250 TS16840-1000
4-Fluoro-3-nitroaniline 99%
4-Fluoro-3-nitroaniline
Formula: C₆H₅FN₂O₂
MW: 156.12 g/mol
Melting Pt: 96…99 °C
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00007833
CAS Number: 364-76-1
EINECS: 206-665-5

Order Now

Specification Test Results

Appearance Ochre Powder
Infrared spectrum Conforms
Melting point 96 to 99 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR