4-Acetylbenzonitrile 98+%

Supplier: Thermo Scientific Chemicals
Warning

16758-0050 16758-0250 16758-1000
TS16758-0050EA 47 USD
TS16758-0050 TS16758-0250 TS16758-1000
4-Acetylbenzonitrile 98+%
4-Acetylbenzonitrile
Formula: C₉H₇NO
MW: 145.16 g/mol
Melting Pt: 55…58 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001825
CAS Number: 1443-80-7
EINECS: 215-885-0

Order Now

Specification Test Results

Appearance Light yellow to beige Crystalline powder or mass and/or chunks
Infrared spectrum Conforms
Melting point 55 to 58 °C
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR