2,6-Pyridinedimethanol 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2,6-Bis(hydroxymethyl)pyridine
16755-0050
16755-0250
16755-1000
TS16755-0050EA
63
USD
TS16755-0050
TS16755-0250
TS16755-1000
2,6-Pyridinedimethanol 98%
2,6-Pyridinedimethanol
Formula:
C₇H₉NO₂ MW: 139.15 g/mol Melting Pt: 112…115 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006351 CAS Number: 1195-59-1 EINECS: 214-803-0 |
Specification Test Results
Appearance | White to yellow to beige Powder |
Infrared spectrum | Conforms |
Melting point | 108 to 115 °C |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...