4-Methylcyclohexanol cis- and trans-mixture 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
16690-0010
16690-2500
TS16690-0010EA
223
USD
TS16690-0010
TS16690-2500
4-Methylcyclohexanol cis- and trans-mixture 99%
4-Methylcyclohexanol
Formula:
C₇H₁₄O Boiling Pt: 171 °C (1013 hPa) Melting Pt: –41 °C Density: 0.92 g/cm³ (0 °C) Flash Pt: 68 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001449 CAS Number: 589-91-3 EINECS: 209-664-8 |
Specification Test Results
Appearance | Clear colorless to yellow Viscous liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4570 to 1.4590 (20 °C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...