4-Piperidone monohydrate hydrochloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4,4-Piperidinediol Hydrochloride, 4,4-Dihydroxypiperidine Hydrochloride, 4-Ketopiperidine Monohydrate Hydrochloride, 4-Piperidinone monohydrate hydrochloride, Piperidin-4-one hydrate hydrochloride, 4-Azacyclohexanone Monohydrate Hydrochloride
16619-0500
16619-2500
TS16619-0500EA
124
USD
TS16619-0500
TS16619-2500
4-Piperidone monohydrate hydrochloride 98%
4-Piperidone monohydrate hydrochloride
Formula:
C₅H₉NO·xHCl·yH₂O Melting Pt: 89…99 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00012776 CAS Number: 40064-34-4 |
Specification Test Results
Appearance | Off-white to yellow to light beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 89 to 99 °C |
Water | 10.5 to 13 % |
TLC | ≥97.5 % |
Titration with HClO4 | ≥97.5 % (On anhydrous substance) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...