(±)-4-Hydroxy-3-methoxymandelic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Hydroxy-3-methoxy-DL-mandelic acid, VMA, Vanillyl mandelic acid, DL-4-Hydroxy-3-methoxymandelic acid, (±)-4-Hydroxy-3-methoxyphenylglycolic acid, 2-Hydroxy-2-(4-hydroxy-3-methoxyphenyl)acetic acid, Vanillylmandelic acid

16490-0010 16490-0050 16490-0250
TS16490-0010EA 61 USD
TS16490-0010 TS16490-0050 TS16490-0250
(±)-4-Hydroxy-3-methoxymandelic acid 99%
(±)-4-Hydroxy-3-methoxymandelic acid
Formula: C₉H₁₀O₅
MW: 198.18 g/mol
Melting Pt: 131…134 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00004235
CAS Number: 55-10-7
EINECS: 200-224-0
Merck Index: 12,10070

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 131 to 134 °C
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR