2-Methoxynaphthalene 98%
Supplier: Thermo Scientific Chemicals
16475-0050
16475-5000
TS16475-0050EA
22
USD
TS16475-0050
TS16475-5000
2-Methoxynaphthalene 98%
2-Methoxynaphthalene
Formula:
C₁₁H₁₀O Boiling Pt: 274 °C (1013 hPa) Melting Pt: 73…75 °C |
MDL Number:
MFCD00004061 CAS Number: 93-04-9 EINECS: 202-213-6 Merck Index: 12,06076 |
Specification Test Results
Appearance | White to yellow-brown Crystalline solid |
Infrared spectrum | Conforms |
Melting point | ≥71.5 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...