1,1,1-Tris(hydroxymethyl)propane 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 1,1,1-Trimethylolpropane, Propylidynetrimethanol, Trimethylol propane, Propylidynetris(methanol), 2-Ethyl-2-(hydroxymethyl)-1,3-propanediol

16465-0010 16465-0025 16465-0100 16465-5000
TS16465-0010EA 47 USD
TS16465-0010 TS16465-0025 TS16465-0100 TS16465-5000
1,1,1-Tris(hydroxymethyl)propane 98%
1,1,1-Tris(hydroxymethyl)propane
Formula: C₆H₁₄O₃
MW: 134.18 g/mol
Boiling Pt: 296
Melting Pt: 53
Flash Pt: 172 °C
Storage Temperature: Ambient
MDL Number: MFCD00004694
CAS Number: 77-99-6
EINECS: 201-074-9

Order Now

Specification Test Results

Appearance White Flakes
Infrared spectrum Conforms
Melting point ≥57 °C
Water ≤0.1 %
Ash ≤0.0015 %
Acidity ≤0.002 % (as Formic acid)
Hydroxyl value ≥1235 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR