DL-Camphor 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: DL-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone(±)-1,7,7-Trimethyl-2-norbornanone1,7,7-Trimethylbicyclo[2.2.1]heptan-2-oneCamphor(±)-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone

16464-0010 16464-0051 16464-2500
TS16464-0051EA 219 USD
TS16464-0051 TS16464-0010 TS16464-2500
DL-Camphor 96%
DL-Camphor
Formula: C₁₀H₁₆O
MW: 152.24 g/mol
Boiling Pt: 204 °C (760 mmHg)
Melting Pt: 175…177 °C
Flash Pt: 64 °C
Storage Temperature: Ambient
MDL Number: MFCD00074738
CAS Number: 76-22-2
EINECS: 200-945-0
UN: 2717
ADR: 4.1,III

Order Now

Specification Test Results

Appearance White Crystals and/or chunks
Infrared spectrum Conforms
GC ≥95 %
Residue after evaporation ≤0.1 %
Angle of optical rotation -0.15° to +0.15° (20°C, 589 nm) (c=10, alcohol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR