2,4-Dichlorobenzaldehyde 98%

Supplier: Thermo Scientific Chemicals
Danger

16414-0050 16414-1000 16414-5000
TS16414-0050EA 25 USD
TS16414-0050 TS16414-1000 TS16414-5000
2,4-Dichlorobenzaldehyde 98%
2,4-Dichlorobenzaldehyde
Formula: C₇H₄Cl₂O
Boiling Pt: 233 °C (1013 hPa)
Melting Pt: 69…73 °C
Flash Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00003305
CAS Number: 874-42-0
EINECS: 212-861-1
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance Light yellow to beige Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 68 to 73 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR