5-Acetoxymethylfurfural 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (5-Formylfuran-2-yl)methyl acetate, 5-Formylfurfuryl Acetate, 5-Acetoxymethyl-2-furaldehyde, Acetic Acid 5-Formylfurfuryl Ester

16340-0020 16340-0100
TS16340-0020EA 49 USD
TS16340-0020 TS16340-0100
5-Acetoxymethylfurfural 97%
5-Acetoxymethylfurfural
Formula: C₈H₈O₄
MW: 168.15 g/mol
Melting Pt: 52…56 °C
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00003233
CAS Number: 10551-58-3

Order Now

Specification Test Results

Appearance White to beige to brown Powder or crystals and/or chunks or fused solid
Infrared spectrum Conforms
Melting point 52 to 56 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR