4-Hydroxybenzaldehyde 99%

Supplier: Thermo Scientific Chemicals
Warning

16277-0010 16277-0025 16277-0500 16277-2500
TS16277-0010EA 335 USD
TS16277-0010 TS16277-0025 TS16277-0500 TS16277-2500
4-Hydroxybenzaldehyde 99%
4-Hydroxybenzaldehyde
Formula: C₇H₆O₂
MW: 122.12 g/mol
Melting Pt: 114…118 °C
Storage Temperature: Ambient
MDL Number: MFCD00006939
CAS Number: 123-08-0
EINECS: 204-599-1
Merck Index: 13,04836

Order Now

Specification Test Results

Appearance Yellow to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 114 to 118 °C
GC ≥98.5 %
Stabilizer 2 to 5 ppm BHT
Residue after evaporation ≤0.001 %
Acidity (CH3COOH) ≤0.002
Peroxides ≤0.005 % (as H2O2)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR