Veratraldehyde 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,4-Dimethoxybenzaldehyde

16261-0025 16261-0050 16261-1000 16261-5000
TS16261-0025EA 425 USD
TS16261-0025 TS16261-0050 TS16261-1000 TS16261-5000
Veratraldehyde 99+%
Veratraldehyde
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 281 °C (760 mmHg)
Melting Pt: 42…45 °C
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00003363
CAS Number: 120-14-9
EINECS: 204-373-2
Merck Index: 12,10084

Order Now

Specification Test Results

Appearance White to yellow Lumps or fused solid
Infrared spectrum Conforms
Melting point 42 to 45 °C
Water ≤1 % (K.F.)
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR