Pivalic anhydride 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trimethylacetic anhydride, 2,2-Dimethylpropionic anhydride

16244-0010 16244-0500 16244-2500
TS16244-0010EA 951 USD
TS16244-0010 TS16244-0500 TS16244-2500
Pivalic anhydride 99%
Pivalic anhydride
Formula: C₁₀H₁₈O₃
Boiling Pt: 193 °C (760 mmHg)
Density: 0.91 g/cm³
Flash Pt: 57 °C
Storage Temperature: Ambient
MDL Number: MFCD00008842
CAS Number: 1538-75-6
EINECS: 216-263-1
UN: 2920
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow liquid Liquid
Infrared spectrum Conforms
Refractive index 1.4080 to 1.4100 (20 °C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR