Diglycolic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,2'-Oxydiacetic acid

16226-0025 16226-1000 16226-5000
TS16226-0025EA 1146 USD
TS16226-0025 TS16226-1000 TS16226-5000
Diglycolic acid 98%
Diglycolic acid
Formula: C₄H₆O₅
MW: 134.09 g/mol
Melting Pt: 141…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00004309
CAS Number: 110-99-6
EINECS: 203-823-5

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 141 to 145 °C
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR