Phenylglyoxal monohydrate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,2-Dihydroxy-1-phenylethan-1-one, Benzoylformaldehyde, Phenylglyoxal hydrate, 2,2-Dihydroxyacetophenone

16197-0050 16197-0250 16197-1000
TS16197-0050EA 89 USD
TS16197-0050 TS16197-0250 TS16197-1000
Phenylglyoxal monohydrate 97%
Phenylglyoxal monohydrate
Formula: C₈H₆O₂·H₂O
Boiling Pt: 142 °C (125 mmHg)
Melting Pt: 71…76 °C
Storage Temperature: Ambient
MDL Number: MFCD00149499
CAS Number: 1075-06-5

Order Now

Specification Test Results

Appearance White to light yellow to pink Powder
Infrared spectrum Conforms
Melting point 71 to 76 °C
Titration excess NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR