2-Amino-4-methoxybenzothiazole 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Methoxybenzothiazol-2-ylamine

16035-0010 16035-0050 160350250
TS16035-0010EA 48 USD
TS16035-0010 TS16035-0050 TS160350250
2-Amino-4-methoxybenzothiazole 97%
2-Amino-4-methoxybenzothiazole
Formula: C₈H₈N₂OS
MW: 180.23 g/mol
Melting Pt: 153…155 °C
Storage Temperature: Ambient
MDL Number: MFCD00005792
CAS Number: 5464-79-9

Order Now

Specification Test Results

Appearance Beige Crystalline powder
Infrared spectrum Conforms
Melting point 150 to 153 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR