5-Methyl-2-furaldehyde 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: 5-Methylfurfural, 5-Methyl-2-Furaldehyde

15992-0250 15992-1000
TS15992-0250EA 52 USD
TS15992-0250 TS15992-1000
5-Methyl-2-furaldehyde 98+%
5-Methyl-2-furaldehyde
Formula: C₆H₆O₂
MW: 110.11 g/mol
Boiling Pt: 89 °C (1013 hPa)
Density: 1.1 g/cm³
Flash Pt: 72 °C
MDL Number: MFCD00003232
CAS Number: 620-02-0
EINECS: 210-622-6

Order Now

Specification Test Results

Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5290 to 1.5320 (20 °C, 589 nm)
GC ≥98.0 %
Acid value ≤10 mg KOH/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR