5-Bromosalicylaldehyde 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Bromo-2-hydroxybenzaldehyde

15991-0250 15991-1000
TS15991-0250EA 62 USD
TS15991-0250 TS15991-1000
5-Bromosalicylaldehyde 99%
5-Bromosalicylaldehyde
Formula: C₇H₅BrO₂
Melting Pt: 103…107 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00003330
CAS Number: 1761-61-1
EINECS: 217-167-2

Order Now

Specification Test Results

Appearance White to yellow to light beige Powder
Infrared spectrum Conforms
Melting point 103 to 107 °C
GC ≥98.5 %
Residue after evaporation ≤0.001 %
Acidity ≤0.0002 meq/g
Filt.over 0.2 micron PFTE Filtered
Peroxides ≤0.002 % (as H2O2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR