5-Nonanone 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Nonan-5-one

15976-0250 15976-1000
TS15976-0250EA 29 USD
TS15976-0250 TS15976-1000
5-Nonanone 98%
5-Nonanone
Formula: C₉H₁₈O
Boiling Pt: 187 °C (1013 hPa)
Melting Pt: –50 °C
Density: 0.82 g/cm³
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00009484
CAS Number: 502-56-7
EINECS: 207-946-5

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4180 to 1.4200 (20 °C, 589 nm)
GC ≥97.5 %
Residue after evaporation ≤0.001 %
Filt.over 0.2 micron PFTE Filtered
Acidity (CH3COOH) ≤0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR