4,4-Dimethyl-2-pentanone 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4,4-Dimethylpentan-2-one
15971-0050
15971-0250
159711000
TS15971-0050EA
151
USD
TS15971-0050
TS15971-0250
TS159711000
4,4-Dimethyl-2-pentanone 99%
4,4-Dimethyl-2-pentanone
Formula:
C₇H₁₄O MW: 114.19 g/mol Boiling Pt: 125…130 °C (760 mmHg) Melting Pt: –64 °C Density: 0.804 g/cm³ Flash Pt: 18 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008854 CAS Number: 590-50-1 EINECS: 209-685-2 UN: 1224 ADR: 3,II |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4025 to 1.4045 (20 °C, 589 nm) |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...