Succinic acid 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,4-Butanedioic acid

15874-0010 15874-0025 15874-0100 15874-2500 15874-5000
TS15874-0010EA 116 USD
TS15874-0010 TS15874-0025 TS15874-0100 TS15874-2500 TS15874-5000
Succinic acid 99%
Succinic acid
Formula: HOOCCH₂CH₂COOH
MW: 118.09 g/mol
Boiling Pt: 235
Melting Pt: 182
Flash Pt: 206 °C
Storage Temperature: Ambient
MDL Number: MFCD00002789
CAS Number: 110-15-6
EINECS: 203-740-4
Merck Index: 13,08953

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 185 to 190 °C
Water ≤1 % (K.F.)
Titration with NaOH ≥98.5 %
Fumaric and maleic acid ≤1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR