trans-2-Hexenal 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (E)-2-Hexenal, trans-2-Hexenal

15813-0250 15813-1000
TS15813-0250EA 45 USD
TS15813-0250 TS15813-1000
trans-2-Hexenal 99%
trans-2-Hexenal
Formula: C₆H₁₀O
Boiling Pt: 147 °C (1013 hPa)
Density: 0.846 g/cm³
Flash Pt: 35 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007008
CAS Number: 6728-26-3
EINECS: 229-778-1
UN: 1988
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4440 to 1.4500 (20 °C, 589 nm)
Density (20°C) 0.843 to 0.849 g/ml
GC ≥98.5 %
Acid value ≤5 mg KOH/g
Chloride (Cl) ≤5 ppm
Nitrate (NO3) ≤10 ppm
Phosphate (PO4) ≤5 ppm
Arsenic (As) ≤1 ppm
Cadmium (Cd) ≤1 ppm
Calcium (Ca) ≤10 ppm
Copper (Cu) ≤1 ppm
Iron (Fe) ≤2 ppm
Zinc (Zn) ≤1 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR