Tetramethyl orthocarbonate 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tetramethylorthocarbonate

15811-0050 15811-0250
TS15811-0050EA 135 USD
TS15811-0050 TS15811-0250
Tetramethyl orthocarbonate 99%
Tetramethyl orthocarbonate
Formula: C₅H₁₂O₄
MW: 136.15 g/mol
Boiling Pt: 114 °C (760 mmHg)
Melting Pt: –5 °C
Density: 1.02 g/cm³
Flash Pt: 22 °C
Storage Temperature: Ambient
MDL Number: MFCD00008473
CAS Number: 1850-14-2
UN: 3272
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥98.5 %
UV (0.5 M in water)
pH 6.0 to 9.0 (5 % in water)
Chloride and Bromide ≤0.01 % (as Cl)
Iodate (IO3) ≤10 ppm
Sulfate (SO4) ≤10 ppm
Calcium (Ca) ≤10 ppm
Copper (Cu) ≤5 ppm
Iron (Fe) ≤5 ppm
Magnesium (Mg) ≤10 ppm
Sodium (Na) ≤50 ppm
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.005 % (in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR