1,2-Phenylenediacetic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: o-Phenylenediacetic acid, 2,2'-(1,2-Phenylene)diacetic acid

15755-0050 15755-0250
TS15755-0050EA 37 USD
TS15755-0050 TS15755-0250
1,2-Phenylenediacetic acid 99%
1,2-Phenylenediacetic acid
Formula: C₁₀H₁₀O₄
MW: 194.19 g/mol
Melting Pt: 150…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00004329
CAS Number: 7500-53-0
EINECS: 231-355-1

Order Now

Specification Test Results

Appearance Beige to light brown Crystalline powder or needles
Infrared spectrum Conforms
Melting point 150 to 152 °C
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR