2-Methyl-3-butyne-2-ol 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Hydroxy-3-methyl-1-butyne, 2-Methylbut-3-yn-2-ol

15682-0010 15682-0025 15682-2500
TS15682-0010EA 102 USD
TS15682-0010 TS15682-0025 TS15682-2500
2-Methyl-3-butyne-2-ol 98%
2-Methyl-3-butyne-2-ol
Formula: C₅H₈O
MW: 84.12 g/mol
Boiling Pt: 104 °C (760 mmHg)
Melting Pt: 3 °C
Density: 0.8608 g/cm³
Flash Pt: 21 °C
Storage Temperature: Ambient
MDL Number: MFCD00004467
CAS Number: 115-19-5
EINECS: 204-070-5
UN: 1987
ADR: 3,II
Merck Index: 12,06113

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4190 to 1.4220 (20 °C, 589 nm)
GC ≤1.0 % tetrahydrofuran
Color scale ≤100 APHA
Iron (Fe) ≤0.001 %
Magnesium (Mg) ≤0.05 %
Potassium (K) ≤0.01 %
Sodium (Na) ≤0.02 %
Strontium (Sr) ≤0.05 %
Heavy metals (as Pb) ≤0.005 %
Titrable acid ≤0.035 meq/g
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR