1,1,3,3-Tetraethoxypropane 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Malonaldehyde Bis(diethyl Acetal)

15673-0050 15673-1000 15673-5000
TS15673-0050EA 25 USD
TS15673-0050 TS15673-1000 TS15673-5000
1,1,3,3-Tetraethoxypropane 97%
1,1,3,3-Tetraethoxypropane
Formula: C₁₁H₂₄O₄
MW: 220.31 g/mol
Boiling Pt: 220 °C (1013 hPa)
Melting Pt: –90 °C
Density: 0.91 g/cm³
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00009240
CAS Number: 122-31-6
EINECS: 204-533-1

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4100 to 1.4120 (20 °C, 589 nm)
GC ≥96.0 %
Sulfate (SO4) ≤0.01 %
Sodium carbonate (Na2CO3) ≤0.2 %
Ammonium (NH4) ≤0.002 %
Iron (Fe) ≤0.0002 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 % (in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR