3-Methyl-3-buten-1-ol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Methylbut-3-en-1-ol, 2-Methyl-1-buten-4-ol

15663-1000 15663-5000
TS15663-1000EA 44 USD
TS15663-1000 TS15663-5000
3-Methyl-3-buten-1-ol 97%
3-Methyl-3-buten-1-ol
Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 132 °C (1013 hPa)
Melting Pt: –82 °C
Density: 0.85 g/cm³
Flash Pt: 36 °C
Storage Temperature: Ambient
MDL Number: MFCD00002933
CAS Number: 763-32-6
EINECS: 212-110-8
UN: 1987
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4327 to 1.4347 (20 °C, 589 nm)
GC ≥96.0 %
Titration with HClO4 ≥98.5 %
Insoluble matter ≤0.0005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR