Ethyl diethoxyacetate 98%

Supplier: Thermo Scientific Chemicals

15607-0250 15607-1000
TS15607-0250EA 72 USD
TS15607-0250 TS15607-1000
Ethyl diethoxyacetate 98%
Ethyl diethoxyacetate
Formula: C₈H₁₆O₄
Boiling Pt: 198…200 °C (761 mmHg)
Density: 0.989 g/cm³
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00009231
CAS Number: 6065-82-3

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4100 to 1.4120 (20 °C, 589 nm)
GC ≥97.5 %
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Sulfur compounds (as S) ≤0.005 %
Water soluble titr. acid ≤0.0003 meg/g
Thiophene Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR