Ethyl-2,2-dimethyl-3-(2-methyl-1-propenyl)cyclopropane-1-carboxylate cis- and trans-mixture 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Ethyl Chrysanthemate, 2,2-Dimethyl-3-(2-methyl-1-propenyl)cyclopropane-1-carboxylic Acid Ethyl Ester, Chrysanthemylmonocarboxylic Acid Ethyl Ester, Ethyl Chrysanthemumate

15606-0250 15606-1000 15606-5000
TS15606-0250EA 34 USD
TS15606-0250 TS15606-1000 TS15606-5000
Ethyl-2,2-dimethyl-3-(2-methyl-1-propenyl)cyclopropane-1-carboxylate cis- and trans-mixture 95%
Ethyl-2,2-dimethyl-3-(2-methyl-1-propenyl)cyclopropane-1-carboxylate
Formula: C₁₂H₂₀O₂
MW: 196.29 g/mol
Boiling Pt: 112 °C (10 mmHg)
Density: 0.9 g/cm³
Flash Pt: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD00001304
CAS Number: 97-41-6

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4590 to 1.4610 (20 °C, 589 nm)
Water ≤0.2 %
GC ≥94 %
Residue after evaporation ≤0.001 %
Carbonyl compounds ≤0.03 % (as C2H5CHO)
Isopropyl alcohol ≤0.05 %
Titrable acid ≤0.0004 meq/g
E/E ≤0.01 %
Methanol ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR