Di(2-pyridyl) ketone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Di(pyridin-2-yl)methanone

15567-0050 15567-0250
TS15567-0050EA 108 USD
TS15567-0050 TS15567-0250
Di(2-pyridyl) ketone 98%
Di(2-pyridyl) ketone
Formula: C₁₁H₈N₂O
MW: 184.20 g/mol
Melting Pt: 53…55 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00006288
CAS Number: 19437-26-4
EINECS: 243-067-3

Order Now

Specification Test Results

Appearance Light yellow to beige to brown Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 49 to 54 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR