3,5-Dimethoxybenzaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

15522-0100 15522-0500
TS15522-0100EA 124 USD
TS15522-0100 TS15522-0500
3,5-Dimethoxybenzaldehyde 98%
3,5-Dimethoxybenzaldehyde
Formula: C₉H₁₀O₃
Boiling Pt: 151 °C (16 mmHg)
Melting Pt: 44…48 °C
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00003366
CAS Number: 7311-34-4
EINECS: 230-772-6

Order Now

Specification Test Results

Appearance White to beige Crystalline solid
Infrared spectrum Conforms
Melting point 44 to 48 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR