1,4-Diisopropylbenzene 99%

Supplier: Thermo Scientific Chemicals
Warning

15521-0050 15521-0250 15521-1000
TS15521-0050EA 22 USD
TS15521-0050 TS15521-0250 TS15521-1000
1,4-Diisopropylbenzene 99%
1,4-Diisopropylbenzene
Formula: C₁₂H₁₈
MW: 162.27
Boiling Pt: 203
Melting Pt: -40
Density: 0.86 g/cm³
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00008892
CAS Number: 100-18-5
EINECS: 202-826-9

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4875 to 1.4905 (20 °C, 589 nm)
GC ≥98.5 %
Chloride (Cl) 18 to 19 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR