(R)-(-)-Carvone 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (R)-2-Methyl-5-(prop-1-en-2-yl)cyclohex-2-enone, L-p-Mentha-1(6),8-dien-2-one, (R)-(-)-2-Methyl-5-(1-methylvinyl)cyclohex-2-en-1-one, L(-)-Carvone

15459-0050 15459-1000 15459-5000
TS15459-0050EA 26 USD
TS15459-0050 TS15459-1000 TS15459-5000
(R)-(-)-Carvone 99%
(R)-(-)-Carvone
Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Boiling Pt: 227 °C (1013 hPa)
Melting Pt: 25.2 °C
Density: 0.958 g/cm³
Flash Pt: 88 °C
MDL Number: MFCD00001578
CAS Number: 6485-40-1
EINECS: 229-352-5

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4950 to 1.5010 (20 °C, 589 nm)
GC ≥98.5 %
Specific gravity (25/25°C) 0.956 to 0.961
Specific optical rotation -57° to -62° (20°C, 589 nm) (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR