4-Formylbenzoic acid 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Terephthalaldehydic acid, 4-Carboxybenzaldehyde

15458-0050 15458-0250 15458-1000
TS15458-0050EA 44 USD
TS15458-0050 TS15458-0250 TS15458-1000
4-Formylbenzoic acid 96%
4-Formylbenzoic acid
Formula: C₈H₆O₃
MW: 150.13 g/mol
Melting Pt: 247 °C
Storage Temperature: Ambient
MDL Number: MFCD00006951
CAS Number: 619-66-9
EINECS: 210-607-4

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
HPLC ≥95 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR