(±)-Camphorquinone 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 1,7,7-Trimethylbicyclo[2.2.1]heptane-2,3-dione, DL-Camphoroquinone

15457-0050 15457-0100 15457-0500
TS15457-0050EA 72 USD
TS15457-0050 TS15457-0100 TS15457-0500
(±)-Camphorquinone 99%
(±)-Camphorquinone
Formula: C₁₀H₁₄O₂
MW: 166.22 g/mol
Melting Pt: 198…202 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064160
CAS Number: 10373-78-1
EINECS: 233-814-1

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 199 to 204 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR