Butyrophenone 99%

Supplier: Thermo Scientific Chemicals

15441-1000 15441-5000
TS15441-1000EA 107 USD
TS15441-1000 TS15441-5000
Butyrophenone 99%
Butyrophenone
Formula: C₆H₅C(O)(CH₂)₂CH₃
Boiling Pt: 228…230 °C (760 mmHg)
Melting Pt: 11…13 °C
Density: 0.988 g/cm³
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00009397
CAS Number: 495-40-9
EINECS: 207-799-7

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5185 to 1.5205 (20 °C, 589 nm)
GC ≥98.5 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR