(±)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(2,2-Dimethyl-1,3-dioxolan-4-yl)methanol, 2,2-Dimethyl-1,3-dioxolan-4-ylmethanol, DL-1,2-Isopropylidenglycerol, 1,2-O-Isopropylideneglycerol, Solketal, (±)-1,2-O-Isopropylidenglycerol, DL-1,2-Isopropylideneglycerol
15365-0025
15365-1000
15365-5000
TS15365-0025EA
266
USD
TS15365-0025
TS15365-1000
TS15365-5000
(±)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol 97%
(±)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol
Formula:
C₆H₁₂O₃ MW: 132.16 g/mol Boiling Pt: 188…189 °C (760 mmHg) Melting Pt: –50 °C Density: 1.06 g/cm³ Flash Pt: 90 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00063238 CAS Number: 100-79-8 EINECS: 202-888-7 Merck Index: 13,05233 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4320 to 1.4360 (20 °C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...