Chroman-4-one 96%
Supplier: Thermo Scientific Chemicals
Synonyms:
4-Chromanone
15346-0100
15346-0500
TS15346-0100EA
114
USD
TS15346-0100
TS15346-0500
Chroman-4-one 96%
Chroman-4-one
Formula:
C₉H₈O₂ MW: 148.16 g/mol Boiling Pt: 253 Melting Pt: 39 Flash Pt: 110 °C |
MDL Number:
MFCD00006840 CAS Number: 491-37-2 EINECS: 207-736-3 |
Specification Test Results
Appearance | White to yellow Crystalline powder or crystals or low melting mass |
Infrared spectrum | Conforms |
Melting point | 34 to 40 °C |
GC | ≥95 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...