4-Methoxy-3-buten-2-one 90% tech.

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Methoxybut-3-en-2-one

15339-0100 15339-0500
TS15339-0100EA 57 USD
TS15339-0100 TS15339-0500
4-Methoxy-3-buten-2-one 90% tech.
4-Methoxy-3-buten-2-one
Formula: C₅H₈O₂
MW: 100.117 g/mol
Boiling Pt: 200 °C (760 mmHg)
Density: 0.98 g/cm³
Flash Pt: 63 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008778
CAS Number: 4652-27-1

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
GC ≥88 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR