Aminoacetaldehyde dimethyl acetal 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,2-Dimethoxyethanamine

15334-0250 15334-1000 15334-5000 153340025
TS15334-0250EA 56 USD
TS15334-0250 TS15334-1000 TS15334-5000 TS153340025
Aminoacetaldehyde dimethyl acetal 99%
Aminoacetaldehyde dimethyl acetal
Formula: C₄H₁₁NO₂
Boiling Pt: 135…139 °C (95 mmHg)
Density: 0.97 g/cm³
Flash Pt: 41 °C
Storage Temperature: Ambient
MDL Number: MFCD00008135
CAS Number: 22483-09-6
EINECS: 245-026-5
UN: 2733
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4160 to 1.4180 (20 °C, 589 nm)
Water ≤0.5 %
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR