2-Hydroxy-m-anisaldehyde 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: o-Vanillin, 2-Hydroxy-3-methoxybenzaldehyde

15307-0100 15307-1000 15307-5000
TS15307-0100EA 39 USD
TS15307-0100 TS15307-1000 TS15307-5000
2-Hydroxy-m-anisaldehyde 99%
2-Hydroxy-m-anisaldehyde
Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 265
Melting Pt: 40…42 °C
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00003322
CAS Number: 148-53-8
EINECS: 205-715-3

Order Now

Specification Test Results

Appearance Yellow to brown Crystals or flakes or crystalline mass
Infrared spectrum Conforms
Melting point 42 to 45 °C
GC ≥98.5 %
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR