4'-Morpholinoacetophenone 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-[4-(Morpholin-4-yl)phenyl]ethan-1-one

15290-0050 15290-0500
TS15290-0050EA 44 USD
TS15290-0050 TS15290-0500
4'-Morpholinoacetophenone 99%
4'-Morpholinoacetophenone
Formula: C₁₂H₁₅NO₂
MW: 205.2530 g/mol
Melting Pt: 94…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00006168
CAS Number: 39910-98-0
EINECS: 254-698-9

Order Now

Specification Test Results

Appearance Yellow to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 94 to 98 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR