2,6-Dichlorophenolindophenol sodium salt hydrate 98+%
Supplier: Thermo Scientific Chemicals
15287-0010
15287-0100
15287-0250
15287-1000
TS15287-0010EA
33
USD
TS15287-0010
TS15287-0100
TS15287-0250
TS15287-1000
2,6-Dichlorophenolindophenol sodium salt hydrate 98+%
2,6-Dichlorophenolindophenol sodium salt hydrate
Formula:
C₁₂H₆Cl₂NNaO₂·xH₂O Storage Temperature: Ambient |
MDL Number:
MFCD00150014 CAS Number: 1266615-56-8 EINECS: 210-640-4 |
Specification Test Results
Appearance | Dark green to dark blue-purple Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤14 % (1 g, 120 °C) |
Titration with HClO4 | ≥98.0 % (On dry substance) |
Suitability | (for Ascorbic acid det.) Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...