9-Decen-1-ol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Decen-10-ol

15264-0050 15264-0250 15264-1000
TS15264-0050EA 25 USD
TS15264-0050 TS15264-0250 TS15264-1000
9-Decen-1-ol 99%
9-Decen-1-ol
Formula: C₁₀H₂₀O
MW: 156.27 g/mol
Boiling Pt: 234…238 °C (760 mmHg)
Melting Pt: –13 °C
Density: 0.845 g/cm³
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00002992
CAS Number: 13019-22-2
EINECS: 235-878-6

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥92 % (main isomer)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR